438170

Sigma-Aldrich

Poly(ethylene glycol-ran-propylene glycol) monobutyl ether

average Mn ~1,700

Linear Formula:
CH3(CH2)3(OCH2CH2)x[OCH2CH(CH3)]yOH
CAS Number:
MDL number:
PubChem Substance ID:

mol wt

average Mn ~1,700

refractive index

n20/D 1.458

viscosity

350 cSt(20 °C)(lit.)

bp

>200 °C (lit.)

transition temp

pour point −34 °C

solubility

H2O: soluble at <40 °C

density

1.051 g/mL at 25 °C

SMILES string

OCCO.CCCCO.CC(O)CO

InChI

1S/C4H10O.C3H8O2.C2H6O2/c1-2-3-4-5;1-3(5)2-4;3-1-2-4/h5H,2-4H2,1H3;3-5H,2H2,1H3;3-4H,1-2H2

InChI key

QMNOIORHZMRPLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hydraulic fluids, metal working fluids and lubricants, heat transfer fluids, solder assist fluids, quenchants, lubricants, solvents, plasticizers and foam control agents.

Features and Benefits

Good lubricity under load, noncorrosive to metals. Very low ash content, low pour point, excellent thermal stability.

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

1

Flash Point(F)

235.4 °F

Flash Point(C)

113 °C

Bahareh Yahyaei et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 101, 343-348 (2012-11-06)
Ag nanoparticles have been synthesized via UV irradiation of ethanolic solution of AgNO3 in presence of pluronic F127 surfactant. This study is aimed at developing a rapid, simple and green method to prepare Ag nanoparticles and understanding its generation kinetics....
Supramolecular assembly of cyclodextrin-based nanospheres for gene delivery.
Min-min Fan et al.
Journal of controlled release : official journal of the Controlled Release Society, 152 Suppl 1, e141-e142 (2011-12-27)
J Araújo et al.
International journal of pharmaceutics, 393(1-2), 167-175 (2010-04-07)
The purpose of this study was to develop a novel nanostructured lipid carrier (NLC) for the intravitreal-targeting delivery of triamcinolone acetonide (TA) by direct ocular instillation. A five-level central composite rotable design was used to study the influence of four...
Diego A Chiappetta et al.
Nanomedicine (London, England), 8(2), 223-237 (2012-11-24)
To investigate the intranasal administration of poly(ethylene oxide)-poly(propylene oxide) polymeric micelles loaded with high payloads of the first-line antiretroviral drug efavirenz for targeting to the CNS. The effect of micellar size and composition and drug payload was assessed, employing simple...
Tarek A Ahmed et al.
Pharmaceutical development and technology, 16(5), 497-510 (2010-06-17)
A suitable topical formulation of mefenamic acid was developed in order to eliminate the gastrointestinal disorders associated with its oral administration. Drug coprecipitates prepared with different polymers at various drug-to-polymer ratios improved drug solubility and dissolution compared to pure drug...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.