MilliporeSigma
All Photos(1)

441813

Sigma-Aldrich

5,10,15,20-Tetrakis(4-sulfonatophenyl)-21H,23H-porphine manganese(III) chloride

Dye content 70 %

Sign Into View Organizational & Contract Pricing

Select a Size

Empirical Formula (Hill Notation):
C44H28ClMnN4O12S4
CAS Number:
Molecular Weight:
1023.36
MDL number:
PubChem Substance ID:
NACRES:
NA.23

form

solid

composition

Dye content, 70%

mp

>300 °C (lit.)

λmax

466 nm

SMILES string

OS(=O)(=O)c1ccc(cc1)-c2c3ccc(n3)c(-c4ccc(cc4)S(O)(=O)=O)c5ccc6c(-c7ccc(cc7)S(O)(=O)=O)c8ccc(n8)c(-c9ccc(cc9)S(O)(=O)=O)c%10ccc2n%10[Mn](Cl)n56

InChI

1S/C44H28N4O12S4.ClH.Mn/c49-61(50,51)29-9-1-25(2-10-29)41-33-17-19-35(45-33)42(26-3-11-30(12-4-26)62(52,53)54)37-21-23-39(47-37)44(28-7-15-32(16-8-28)64(58,59)60)40-24-22-38(48-40)43(36-20-18-34(41)46-36)27-5-13-31(14-6-27)63(55,56)57;;/h1-24H,(H,49,50,51)(H,52,53,54)(H,55,56,57)(H,58,59,60);1H;/q-2;;+3/p-1/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-;;

InChI key

WMITXTXHDITCGT-YKKPBKTHSA-M

General description

5,10,15,20-Tetrakis(4-sulfonatophenyl)-21H,23H-porphine manganese(III) chloride is a metal–porphyrin. During luminol chemiluminescence reactions, it can act as a highly sensitive catalyst. On-line chemiluminescence detection with capillary electrophoresis of the product was investigated and the detection limit was determined to be 361 amol and 8.1 × 10–9 mol L–1.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

An improved apparatus for on-line chemiluminescence detection with capillary electrophoresis and detection of metal?porphyrins
Zhang Y and Zhang H
Analytical Communications, 35(9), 293-296 (1998)
Rohan D A Alvares et al.
Scientific reports, 7(1), 15493-15493 (2017-11-16)
Magnetic resonance imaging (MRI) provides superior resolution of anatomical features and the best soft tissue contrast, and is one of the predominant imaging modalities. With this technique, contrast agents are often used to aid discrimination by enhancing specific features. Over

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service