Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

441856

Sigma-Aldrich

Methyl (4S)-(+)-2,2-dimethyl-1,3-dioxolane-4-acetate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O4
CAS Number:
Molecular Weight:
174.19
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

97%

optical activity

[α]20/D +17°, neat

refractive index

n20/D 1.615 (lit.)

bp

192-194 °C (lit.)

density

1.07 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C[C@H]1COC(C)(C)O1

InChI

1S/C8H14O4/c1-8(2)11-5-6(12-8)4-7(9)10-3/h6H,4-5H2,1-3H3/t6-/m0/s1

InChI key

AEMMCWMMNLSHFT-LURJTMIESA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

149.0 °F - closed cup

flash_point_c

65 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service