442607

Sigma-Aldrich

N-(4-Acetylphenyl)-2-chloroacetamide

99%

Linear Formula:
ClCH2CONHC6H4COCH3
CAS Number:
Molecular Weight:
211.64
MDL number:
PubChem Substance ID:

assay

99%

mp

153-155 °C (lit.)

SMILES string

CC(=O)c1ccc(NC(=O)CCl)cc1

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service