Documents

442968

Sigma-Aldrich

trans-1,3-Diphenyl-2,3-epoxypropan-1-one

95%

Synonym(s):
trans-Benzalacetophenone oxide, trans-2-Benzoyl-3-phenyloxirane
Empirical Formula (Hill Notation):
C15H12O2
CAS Number:
Molecular Weight:
224.25
Beilstein/REAXYS Number:
84132
MDL number:

assay

95%

mp

86-89 °C (lit.)

SMILES string

O=C([C@H]1O[C@@H]1c2ccccc2)c3ccccc3

InChI

1S/C15H12O2/c16-13(11-7-3-1-4-8-11)15-14(17-15)12-9-5-2-6-10-12/h1-10,14-15H/t14-,15-/m1/s1

InChI key

UQGMJZQVDNZRKT-HUUCEWRRSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service