Documents

448451

Sigma-Aldrich

(+)-mono-(1S)-Menthyl phthalate

99%

Empirical Formula (Hill Notation):
C18H24O4
CAS Number:
Molecular Weight:
304.38
MDL number:

assay

99%

optical activity

[α]20/D +93°, c = 5 in chloroform

mp

108-113 °C (lit.)

SMILES string

CC(C)[C@H]1CC[C@H](C)C[C@@H]1OC(=O)c2ccccc2C(O)=O

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service