448494

Sigma-Aldrich

Tetrasodium tetrakis(μ-carbonato)dirhodate hydrate

Linear Formula:
Na4[Rh2(CO3)4]·xH2O
Molecular Weight:
537.81 (anhydrous basis)
MDL number:
PubChem Substance ID:

form

powder

mp

>350 °C (lit.)

SMILES string

[Na+].[Na+].[Na+].[Na+].[Rh++][Rh++].[H]O[H].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service