448915

Sigma-Aldrich

Tris(dimethylsilyloxy)methylsilane

97%

Synonym(s):
Tris(dimethylsiloxy)(methyl)silane, Methyl-tris(dimethylsiloxy)silane
Linear Formula:
[(CH3)2SiHO]3SiCH3
CAS Number:
Molecular Weight:
268.61
Beilstein/REAXYS Number:
1764389
EC Number:
MDL number:

assay

97%

refractive index

n20/D 1.384 (lit.)

bp

68 °C/5 mmHg (lit.)

density

0.861 g/mL at 25 °C (lit.)

SMILES string

C[SiH](C)O[Si](C)(O[SiH](C)C)O[SiH](C)C

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service