44911

Sigma-Aldrich

N-Alloc-1,5-pentanediamine hydrochloride

≥98.0% (AT)

Synonym(s):
Allyl N-(5-aminopentyl)carbamate hydrochloride, N-Alloc-1,5-diaminopentane hydrochloride
Empirical Formula (Hill Notation):
C9H18N2O2
CAS Number:
Molecular Weight:
186.25
MDL number:
PubChem Substance ID:

assay

≥98.0% (AT)

reaction suitability

reagent type: cross-linking reagent

SMILES string

Cl[H].NCCCCCNC(=O)OCC=C

InChI

1S/C9H18N2O2.ClH/c1-2-8-13-9(12)11-7-5-3-4-6-10;/h2H,1,3-8,10H2,(H,11,12);1H

InChI key

RXDHRWKGTMXRAD-UHFFFAOYSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service