MilliporeSigma
All Photos(1)

449121

Sigma-Aldrich

Mercury(II) bromide

anhydrous, beads, −10 mesh, 99.999% trace metals basis

Synonym(s):
Mercuric bromide
Linear Formula:
HgBr2
CAS Number:
Molecular Weight:
360.40
EC Number:
MDL number:
PubChem Substance ID:

grade

anhydrous

vapor pressure

1 mmHg ( 136.5 °C)

assay

99.999% trace metals basis

form

beads

particle size

−10 mesh

bp

322 °C (lit.)

mp

236 °C (lit.)

solubility

ethanol: slightly soluble(lit.)
methanol: slightly soluble(lit.)

SMILES string

Br[Hg]Br

InChI

1S/2BrH.Hg/h2*1H;/q;;+2/p-2

InChI key

NGYIMTKLQULBOO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service