Documents

449237

Sigma-Aldrich

Silver perchlorate

99.9%

Linear Formula:
AgClO4
CAS Number:
Molecular Weight:
207.32
EC Number:
MDL number:

assay

99.9%

composition

Ag, >50.5%

density

2.8 g/mL at 25 °C (lit.)

SMILES string

[Ag+].[O-]Cl(=O)(=O)=O

InChI

1S/Ag.ClHO4/c;2-1(3,4)5/h;(H,2,3,4,5)/q+1;/p-1

InChI key

YDHABVNRCBNRNZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Used to prepare silver 3,3′-dicyanodiphenylacetylene coordination networks for study of the effect of ligands on network structure toward the goal of engineering novel materials.

RIDADR

UN 1481 5.1/PG 2

WGK Germany

3

Certificate of Analysis
Certificate of Origin
C Rentel et al.
Analytical chemistry, 70(20), 4394-4400 (1998-10-31)
In LC/MS, nonpolar substances in the majority of cases cannot be ionized by standard electrospray ionization (ESI) because they obviously lack a site for protonation or deprotonation. The ionization of carotenoids and tocopherols can be greatly enhanced by the addition...
E Petráková et al.
Carbohydrate research, 233, 101-112 (1992-09-02)
Specifically deoxygenated methyl alpha-isomaltotriosides were synthesized by the silver perchlorate-mediated condensation of a protected alpha-isomaltosyl chloride with suitably blocked derivatives of methyl alpha-D-glucopyranoside deoxygenated respectively at positions 2, 3, and 4, followed by deprotection.
Masafumi Harada et al.
Journal of colloid and interface science, 339(2), 373-381 (2009-09-08)
Metal particles of silver (Ag) were synthesized by the photoreduction of silver perchlorate (AgClO(4)) in water-in-ionic liquid (ILs) microemulsions consisting of Tween 20, water and ionic liquids. The ILs were tetrafluoroborate anions associated with the cations 1-butyl-3-methylimidazolium tetrafluoroborate ([BMIm][BF(4)]) and...
Ken Sakai et al.
Acta crystallographica. Section B, Structural science, 60(Pt 3), 255-262 (2004-05-19)
The third crystal structure containing the hydroxo-bridged cis-diammineplatinum(II) dimer has been determined for a perchlorate salt of the complex, [Pt(2)(NH(3))(4)(micro-OH)(2)](ClO(4))(2). However, the dinuclear cations in the nitrate and the carbonate salts, [Pt(2)(NH(3))(4)(micro-OH)(2)](NO(3))(2) [Faggiani, Lippert, Lock & Rosenberg (1977). J. Am....
Glycosylation via Cp2ZrCl2/AgClO4-mediated activation of anomeric sulfoxides.
P Wipf et al.
The Journal of organic chemistry, 66(23), 7910-7914 (2001-11-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.