All Photos(1)

451118

Sigma-Aldrich

Gadolinium(III) bromide hydrate

99.99% trace metals basis

Linear Formula:
GdBr3 · xH2O
CAS Number:
Molecular Weight:
396.96 (anhydrous basis)
EC Number:
MDL number:
PubChem Substance ID:

assay

99.99% trace metals basis

form

powder and chunks

mp

186 °C (lit.)

SMILES string

O.Br[Gd](Br)Br

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service