Skip to Content
MilliporeSigma

454591

Poly(propylene-co-tetrafluoroethylene)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
[-CH2CH(CH3)-]x(-CF2CF2-)y
CAS Number:
UNSPSC Code:
12162002
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


composition

fluorine, 57 wt. %

hardness

73 (Shore A)

viscosity

35(121 °C, Mooney, ML 1+10)(lit.)

density

1.55 g/mL at 25 °C (lit.)

SMILES string

CC=C.F\C(F)=C(\F)F





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library