All Photos(1)

456594

Sigma-Aldrich

Poly(ethylene azelate)

average Mw ~55,500, average Mn ~29,000

Synonym(s):
Ethylene glycol-azelaic acid polymer, Poly(ethylene azelate)
Linear Formula:
[OCH2CH2O2C(CH2)7CO]n
CAS Number:
MDL number:
PubChem Substance ID:

mol wt

average Mn ~29,000
average Mw ~55,500

transition temp

Tg −54 °C
Tm 52 °C (DSC at peak)

density

1.183 g/mL at 25 °C (lit.)

SMILES string

OCCO.OC(=O)CCCCCCCC(O)=O

InChI

1S/C9H16O4.C2H6O2/c10-8(11)6-4-2-1-3-5-7-9(12)13;3-1-2-4/h1-7H2,(H,10,11)(H,12,13);3-4H,1-2H2

InChI key

HPXYFPBBKMWIFM-UHFFFAOYSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service