458538

Sigma-Aldrich

(3S-cis)-(+)-Tetrahydro-3,7a-diphenylpyrrolo[2,1-b]oxazol-5(6H)-one

98%

Empirical Formula (Hill Notation):
C18H17NO2
CAS Number:
Molecular Weight:
279.33
MDL number:
PubChem Substance ID:

assay

98%

optical activity

[α]20/D +113°, c = 1 in chloroform

mp

82-85 °C (lit.)

SMILES string

O=C1CC[C@]2(OC[C@@H](N12)c3ccccc3)c4ccccc4

Application

Chiral bicyclic lactams such as this are useful templates for the asymmetric synthesis of a variety of natural and unnatural products. High levels of asymmetric induction have been obtained in cycloadditions, alkylations, conjugate additions, annulations, and dihydroxylations.

Certificate of Analysis

Certificate of Origin

Sandham, D.A. Meyers, A.I.
Journal of the Chemical Society. Chemical Communications, 2511-2511 (1995)
Andres, C.S. Meyers, A.I.
Tetrahedron Letters, 36, 3491-3491 (1995)
A Rapid and Efficient Approach to Chiral, Nonracemic Aza Sugars from Nonsugars. A Formal Synthesis of 1,4-Dideoxy-1,4-imino-D-lyxitol.
A. I. Meyers et al.
The Journal of organic chemistry, 61(8), 2586-2587 (1996-04-19)
Meyers, A.I. et al.
The Journal of Organic Chemistry, 60, 4359-4359 (1995)
Brengel, G.P. Meyers, A.I.
The Journal of Organic Chemistry, 61, 3230-3230 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service