458546

Sigma-Aldrich

(3R-cis)-(−)-2,3-Dihydro-3-isopropyl-7a-methylpyrrolo[2,1-b]oxazol-5(7aH)-one

98%

Empirical Formula (Hill Notation):
C10H15NO2
CAS Number:
Molecular Weight:
181.23
MDL number:
PubChem Substance ID:

assay

98%

optical activity

[α]22/D −37°, c = 1 in chloroform

mp

47-50 °C (lit.)

SMILES string

CC(C)[C@@H]1CO[C@@]2(C)C=CC(=O)N12

InChI

1S/C10H15NO2/c1-7(2)8-6-13-10(3)5-4-9(12)11(8)10/h4-5,7-8H,6H2,1-3H3/t8-,10-/m0/s1

InChI key

JOXQDNAGAGXGOK-WPRPVWTQSA-N

Related Categories

Packaging

500 mg in glass bottle

Application

High levels of asymmetric induction have been obtained with this chiral bicyclic lactam in cycloadditions, alkylations, conjugate additions, annulations, and dihydroxylations.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Munchhoff, M.J. Meyers, A.I.
The Journal of Organic Chemistry, 60, 7084-7084 (1995)
Fray, A.H. Meyers, A.I.
The Journal of Organic Chemistry, 61, 3362-3362 (1996)
Brengel, G.P. Meyers, A.I.
The Journal of Organic Chemistry, 61, 3230-3230 (1996)
Andres, C.J. et al.
Tetrahedron Letters, 60, 3189-3189 (1995)
Munchhoff, M.J. Meters, A.I.
The Journal of Organic Chemistry, 60, 7086-7086 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.