Skip to Content
MilliporeSigma

460508

(S)-(+)-Glycidyl butyrate

98%

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C7H12O3
CAS Number:
Molecular Weight:
144.17
UNSPSC Code:
12352005
PubChem Substance ID:
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI

1S/C7H12O3/c1-2-3-7(8)10-5-6-4-9-6/h6H,2-5H2,1H3/t6-/m0/s1

SMILES string

CCCC(=O)OC[C@@H]1CO1

InChI key

YLNSNVGRSIOCEU-LURJTMIESA-N

assay

98%

optical activity

[α]21/D +31°, neat

refractive index

n20/D 1.428 (lit.)

bp

90 °C/19 mmHg (lit.)

density

1.018 g/mL at 25 °C (lit.)

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

185.0 °F - closed cup

flash_point_c

85 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arnaldo Glogauer et al.
Microbial cell factories, 10, 54-54 (2011-07-19)
Metagenomics, the application of molecular genomics to consortia of non-cultivated microbes, has the potential to have a substantial impact on the search for novel industrial enzymes such as esterases (carboxyl ester hydrolases, EC 3.1.1.1) and lipases (triacylglycerol lipases, EC 3.1.1.3).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service