Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

460559

Sigma-Aldrich

Serotonin hydrochloride

Synonym(s):

3-(2-Aminoethyl)-5-hydroxyindole hydrochloride, 5-HT, 5-Hydroxytryptamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O · HCl
CAS Number:
Molecular Weight:
212.68
Beilstein/REAXYS Number:
3570963
MDL number:
UNSPSC Code:
12352200

impurities

<4% ethanol
<5% water

mp

149-154 °C (lit.)

SMILES string

Cl[H].NCCc1c[nH]c2ccc(O)cc12

InChI

1S/C10H12N2O.ClH/c11-4-3-7-6-12-10-2-1-8(13)5-9(7)10;/h1-2,5-6,12-13H,3-4,11H2;1H

InChI key

MDIGAZPGKJFIAH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Neurotransmitter.

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Repr. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service