461482

Sigma-Aldrich

(S)-(−)-N-Methyl-1-(1-naphthyl)ethylamine

97%

Linear Formula:
C10H7CH(CH3)NHCH3
CAS Number:
Molecular Weight:
185.26
MDL number:
PubChem Substance ID:

assay

97%

optical activity

[α]23/D −89°, c = 4 in ethanol

refractive index

n20/D 1.605 (lit.)

bp

203 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

SMILES string

CN[C@@H](C)c1cccc2ccccc12

InChI

1S/C13H15N/c1-10(14-2)12-9-5-7-11-6-3-4-8-13(11)12/h3-10,14H,1-2H3/t10-/m0/s1

InChI key

YYETYHLXPGYQPZ-JTQLQIEISA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service