461636

Sigma-Aldrich

(R)-(+)-N-[1-(1-Naphthyl)ethyl]succinamic acid

98%

Linear Formula:
C10H7CH(CH3)NHCOCH2CH2CO2H
CAS Number:
Molecular Weight:
271.31
MDL number:
PubChem Substance ID:

assay

98%

optical activity

[α]24/D +45°, c = 2 in ethanol

mp

157-159 °C (lit.)

SMILES string

C[C@@H](NC(=O)CCC(O)=O)c1cccc2ccccc12

InChI

1S/C16H17NO3/c1-11(17-15(18)9-10-16(19)20)13-8-4-6-12-5-2-3-7-14(12)13/h2-8,11H,9-10H2,1H3,(H,17,18)(H,19,20)/t11-/m1/s1

InChI key

AUSXHLAOQYCBAR-LLVKDONJSA-N

Application

Acid for the resolution of amines.

Certificate of Analysis

Certificate of Origin

Kozma, D. et al.
J. Therm. Anal., 44, 339-339 (1995)
Acs, M. et al.
Chirality, 6, 314-314 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service