Documents

464333

Sigma-Aldrich

Poly(styrene-co-divinylbenzene)-graft-poly(ethylene glycol), amine terminated

powder, 150 μm±25 μm bead diameter

form

powder

specific activity

0.4-0.5 meq/g (Amine values)

bead diameter

150 μm±25 μm

Application

Solid phase synthesis support.

Features and Benefits

Swollen by polar and apolar solvents such as water, methanol, DMF and THF.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service