Documents

465771

Sigma-Aldrich

N-tert-Butyl-1,1-dimethyl-1-(2,3,4,5-tetramethyl-2,4-cyclopentadien-1-yl)silanamine

98%

Empirical Formula (Hill Notation):
C15H29NSi
CAS Number:
Molecular Weight:
251.48
MDL number:
PubChem Substance ID:

assay

98%

refractive index

n20/D 1.485 (lit.)

bp

110 °C/3 mmHg (lit.)

density

0.878 g/mL at 25 °C (lit.)

SMILES string

CC1=C(C)C(C)=C(C)C1[Si](C)(C)NC(C)(C)C

InChI

1S/C15H29NSi/c1-10-11(2)13(4)14(12(10)3)17(8,9)16-15(5,6)7/h14,16H,1-9H3

InChI key

ZXPSQIUMSOPNIA-UHFFFAOYSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service