Documents

465828

Sigma-Aldrich

2,5-Bis(trifluoromethyl)benzyl alcohol

99%

Linear Formula:
(CF3)2C6H3CH2OH
CAS Number:
Molecular Weight:
244.13
MDL number:
PubChem Substance ID:

assay

99%

mp

34-36 °C (lit.)

SMILES string

OCc1cc(ccc1C(F)(F)F)C(F)(F)F

RIDADR

UN 1325 4.1/PG 2

WGK Germany

3

Flash Point(F)

199 °F

Flash Point(C)

93 °C

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.