All Photos(1)

467413

Sigma-Aldrich

4-Ethyliodobenzene

97%

Linear Formula:
C2H5C6H4I
CAS Number:
Molecular Weight:
232.06
MDL number:
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.59 (lit.)

bp

112-113 °C/20 mmHg (lit.)

density

1.6 g/mL at 25 °C (lit.)

SMILES string

CCc1ccc(I)cc1

InChI

1S/C8H9I/c1-2-7-3-5-8(9)6-4-7/h3-6H,2H2,1H3

InChI key

OOLSRHZMXAYDFB-UHFFFAOYSA-N

Certificate of Analysis

Certificate of Origin

Reactions of Free Radicals with Olefins. Dehydro Dimer Structures of 4-Vinylcyclohexene. The
Shelton JR and Siuda JF.
The Journal of Organic Chemistry, 31(6), 2028-2030 (1966)
Cycloaddition of Dimethylketene to Olefinic Substrates.
Krapcho AP and Lesser JH.
The Journal of Organic Chemistry, 31(6), 2030-2032 (1966)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service