Documents

468606

Sigma-Aldrich

PSS-(2-(5-Norbornen-2-yl)ethyl)-Heptacyclopentyl substituted

Synonym(s):
Norbornenylethyl-POSS®, 1-[2-(5-Norbornen-2-yl)ethyl]-3,5,7,9,11,13,15-heptacyclopentylpentacyclo[9.5.1.13,9.15,15.17,13]octasiloxane
Empirical Formula (Hill Notation):
C44H76O12Si8
CAS Number:
Molecular Weight:
1021.75
MDL number:
PubChem Substance ID:

SMILES string

C1CCC(C1)[Si]23O[Si]6(CCC4C[C@H]5C[C@@H]4C=C5)O[Si]7(O[Si](O2)(O[Si]8(O[Si](O3)(O[Si](O6)(O[Si](O7)(O8)C9CCCC9)C%10CCCC%10)C%11CCCC%11)C%12CCCC%12)C%13CCCC%13)C%14CCCC%14

Application

Polyhedral oligomeric silsesquioxane (POSS) monomer, useful as model for silica surfaces, and for the formation of novel polymeric materials.

Features and Benefits

Polyhedral oligomeric silsesquioxane (POSS) monomer, useful as model for silica surfaces, and for the formation of novel polymeric materials.

Legal Information

POSS is a registered trademark of Hybrid Plastics, Inc.

Certificate of Analysis

Certificate of Origin

Macromolecules, 28, 8435-8435 (1995)
Journal of the American Chemical Society, 112, 1931-1931 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service