Documents

470589

Sigma-Aldrich

Scandium(III) trifluoromethanesulfonimide

97%

Synonym(s):
1,1,1-Trifluoro-N-[(trifluoromethyl)sulfonyl]methanesulfonamide scandium derivative
Linear Formula:
Sc[N(SO2CF3)2]3
CAS Number:
Molecular Weight:
885.39
MDL number:

assay

97%

mp

129-139 °C (lit.)

SMILES string

FC(F)(F)S(=O)(=O)N([Sc](N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F)N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F

InChI

1S/3C2F6NO4S2.Sc/c3*3-1(4,5)14(10,11)9-15(12,13)2(6,7)8;/q3*-1;+3

InChI key

FUXLYEZEIZAKTL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in ampules

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service