MilliporeSigma
All Photos(2)

470597

Sigma-Aldrich

Z-Tyr-OH

97%

Synonym(s):
N-Carbobenzyloxy-L-tyrosine, Z-L-Tyrosine
Linear Formula:
HOC6H4CH2CH(NHCOOCH2C6H5)COOH
CAS Number:
Molecular Weight:
315.32
Beilstein:
2169918
EC Number:
MDL number:
PubChem Substance ID:

Quality Level

assay

97%

optical activity

[α]22/D +11°, c = 1 in acetic acid

reaction suitability

reaction type: solution phase peptide synthesis

mp

57-60 °C (lit.)

application(s)

peptide synthesis

SMILES string

O=C(N[C@H](C(O)=O)CC1=CC=C(O)C=C1)OCC2=CC=CC=C2

InChI

1S/C17H17NO5.H2O/c19-14-8-6-12(7-9-14)10-15(16(20)21)18-17(22)23-11-13-4-2-1-3-5-13;/h1-9,15,19H,10-11H2,(H,18,22)(H,20,21);1H2/t15-;/m0./s1

InChI key

BBPCQKGWAVVMSL-RSAXXLAASA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Jiae Seo et al.
ACS applied materials & interfaces, 11(30), 27306-27317 (2019-06-27)
A flexible hard coating material displaying extreme scratch resistance and foldable flexibility was developed via the design of an organic-inorganic hybrid coating material employing an alkoxysilyl-functionalized polyrotaxane cross-linker (PRX_Si1). PRX_Si1 has a molecular necklace-like structure that can form organic-inorganic cross-linking

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service