Skip to Content
MilliporeSigma

475173

Tris(cyclopentadienyl)praseodymium(III)

99.9% trace metals basis

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
Pr(C5H5)3
CAS Number:
Molecular Weight:
336.19
UNSPSC Code:
12352300
PubChem Substance ID:
EC Number:
234-298-0
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Tris(cyclopentadienyl)praseodymium(III), 99.9% trace metals basis

SMILES string

[Pr].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2.[CH]3[CH][CH][CH][CH]3

assay

99.9% trace metals basis

form

solid

mp

420 °C (lit.)

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Water-react 2

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service