Skip to Content
MilliporeSigma

475904

Sigma-Aldrich

3,4-Dichlorophenyl isothiocyanate

97%

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

About This Item

Linear Formula:
Cl2C6H3NCS
CAS Number:
Molecular Weight:
204.08
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

assay

97%

refractive index

n20/D 1.68 (lit.)

bp

134-136 °C/7 mmHg (lit.)

density

1.422 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1Cl)N=C=S

InChI

1S/C7H3Cl2NS/c8-6-2-1-5(10-4-11)3-7(6)9/h1-3H

InChI key

OSBIEFWIIINTNJ-UHFFFAOYSA-N

Other Notes

Material may have a small amount of precipitate which will not affect use

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service