All Photos(1)

476242

Sigma-Aldrich

Bis[4-(2-hydroxyethoxy)phenyl] sulfone

95%

Synonym(s):
2,2′-[Sulfonylbis(4,1-phenyleneoxy)]bis[ethanol], 2,2′-((Sulfonylbis(4,1-phenylene))bis(oxy))diethanol, Bisphenol S bis(2-hydroxyethyl) ether, Bis[4-(β-hydroxyethoxy)phenyl] sulfone
Linear Formula:
(HOCH2CH2OC6H4)2SO2
CAS Number:
Molecular Weight:
338.38
EC Number:
MDL number:
PubChem Substance ID:

Quality Level

assay

95%

mp

179-183 °C (lit.)

SMILES string

OCCOc1ccc(cc1)S(=O)(=O)c2ccc(OCCO)cc2

InChI

1S/C16H18O6S/c17-9-11-21-13-1-5-15(6-2-13)23(19,20)16-7-3-14(4-8-16)22-12-10-18/h1-8,17-18H,9-12H2

InChI key

UTNSTOOXQPHXJQ-UHFFFAOYSA-N

Certificate of Analysis

Certificate of Origin

Katra Kolšek et al.
Toxicology in vitro : an international journal published in association with BIBRA, 29(1), 8-15 (2014-09-07)
A homeostasis of the glucocorticoid and androgen endocrine system is essential to human health. Their disturbance can lead to various diseases, for example cardiovascular, inflammatory and autoimmune diseases, infertility, cancer. Fifteen widely used industrial chemicals that disrupt endocrine activity were...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service