MilliporeSigma
All Photos(1)

476765

Sigma-Aldrich

Cobalt(II) fluoride

anhydrous, powder, 99.99%

Synonym(s):
Cobaltous fluoride
Linear Formula:
CoF2
CAS Number:
Molecular Weight:
96.93
EC Number:
MDL number:
PubChem Substance ID:

grade

anhydrous

assay

99.99%

form

powder

density

4.46 g/mL at 25 °C

SMILES string

F[Co]F

InChI

1S/Co.2FH/h;2*1H/q+2;;/p-2

InChI key

YCYBZKSMUPTWEE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service