Documents

477362

Sigma-Aldrich

(R)-(+)-1-Fluoro-2-octanol

97%

Linear Formula:
CH3(CH2)5CH(OH)CH2F
CAS Number:
Molecular Weight:
148.22
MDL number:
PubChem Substance ID:

assay

97%

optical activity

[α]25/D +10°, neat

bp

96-97 °C/22 mmHg (lit.)

density

1.254 g/mL at 25 °C (lit.)

SMILES string

CCCCCC[C@@H](O)CF

InChI

1S/C8H17FO/c1-2-3-4-5-6-8(10)7-9/h8,10H,2-7H2,1H3/t8-/m1/s1

InChI key

KLGNQHRPPLDIFC-MRVPVSSYSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service