Skip to Content
MilliporeSigma

477419

Pentamethylcyclopentadienyltantalum(IV) tetrachloride

97%

Synonym(s):

Tetrachloropentamethylcyclopentadienyltantalum(IV)

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C10H15Cl4Ta
CAS Number:
Molecular Weight:
457.99
UNSPSC Code:
12352300
PubChem Substance ID:
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

SMILES string

Cl[Ta](Cl)(Cl)Cl.C[C]1[C](C)[C](C)[C](C)[C]1C

assay

97%

mp

>300 °C (lit.)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service