All Photos(1)

478601

Sigma-Aldrich

2,6-Diethylphenyl isocyanate

98%

Linear Formula:
(C2H5)2C6H3NCO
CAS Number:
Molecular Weight:
175.23
MDL number:
PubChem Substance ID:

assay

98%

refractive index

n20/D 1.528 (lit.)

bp

115 °C/1 mmHg (lit.)

density

1.014 g/mL at 25 °C (lit.)

SMILES string

CCc1cccc(CC)c1N=C=O

InChI

1S/C11H13NO/c1-3-9-6-5-7-10(4-2)11(9)12-8-13/h5-7H,3-4H2,1-2H3

InChI key

AUSRFKVPUCQCCN-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Certificate of Analysis

Certificate of Origin

Radoslaw Laufer et al.
Bioorganic & medicinal chemistry, 22(17), 4968-4997 (2014-07-22)
TTK kinase was identified by in-house siRNA screen and pursued as a tractable, novel target for cancer treatment. A screening campaign and systematic optimization, supported by computer modeling led to an indazole core with key sulfamoylphenyl and acetamido moieties at...
Jerry O Ebalunode et al.
Bioorganic & medicinal chemistry, 17(14), 5133-5138 (2009-06-13)
Current anesthetics, especially the inhaled ones, have troublesome side effects and may be associated with durable changes in cognition. It is therefore highly desirable to develop novel chemical entities that reduce these effects while preserving or enhancing anesthetic potency. In...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service