All Photos(1)

480533

Sigma-Aldrich

Potassium nitridotrioxoosmate(VIII)

Linear Formula:
K(NOsO3)
CAS Number:
Molecular Weight:
291.33
EC Number:
MDL number:
PubChem Substance ID:

form

crystalline

mp

>187 °C (dec.) (lit.)

density

4.2 g/mL at 25 °C (lit.)

SMILES string

[K]N=[Os](=O)(=O)=O

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service