Skip to Content
MilliporeSigma

481491

Dysprosium(III) acetylacetonate hydrate

99.9%

Synonym(s):

2,4-Pentanedione dysprosium(III) derivative, Dy(acac)3

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
Dy(C5H7O2)3 · xH2O
CAS Number:
Molecular Weight:
459.82 (anhydrous basis)
UNSPSC Code:
12352300
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Dysprosium(III) acetylacetonate hydrate, 99.9%

SMILES string

CC(=O)\C=C(\C)O[Dy](O\C(C)=C/C(C)=O)O\C(C)=C/C(C)=O

assay

99.9%

mp

125-130 °C (lit.)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service