Skip to Content
MilliporeSigma

484563

Germanium(IV) selenide

99.999% trace metals basis

Synonym(s):

Germanium diselenide, Germanium selenide

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
GeSe2
CAS Number:
Molecular Weight:
230.56
UNSPSC Code:
12352300
PubChem Substance ID:
EC Number:
235-063-5
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Germanium(IV) selenide, 99.999% trace metals basis

InChI key

WYDSCDJEAVCHQJ-UHFFFAOYSA-N

InChI

1S/GeSe2/c2-1-3

SMILES string

[Se]=[Ge]=[Se]

assay

99.999% trace metals basis

mp

707 °C (lit.)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service