490075

Sigma-Aldrich

Boc-Lys(Z)-OH-ε-15N

98 atom % 15N

Synonym(s):
L-Lysine-ε-15N1, αN-tBoc, εN-Cbz derivative, Nα-(tert-Butoxycarbonyl)-Nε-(carbobenzyloxy)-L-lysine-ε-15N
Linear Formula:
(Cbz)H15N(CH2)4CH(NH-t-Boc)CO2H
CAS Number:
Molecular Weight:
381.43
MDL number:
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % 15N

application(s)

bio NMR: suitable
protein expression: suitable

mp

76-79 °C (lit.)

mass shift

M+1

SMILES string

CC(C)(C)OC(=O)N[C@@H](CCCC[15NH]C(=O)OCc1ccccc1)C(O)=O

InChI

1S/C19H28N2O6/c1-19(2,3)27-18(25)21-15(16(22)23)11-7-8-12-20-17(24)26-13-14-9-5-4-6-10-14/h4-6,9-10,15H,7-8,11-13H2,1-3H3,(H,20,24)(H,21,25)(H,22,23)/t15-/m0/s1/i20+1

InChI key

BDHUTRNYBGWPBL-QYYFNWSZSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service