MilliporeSigma
All Photos(1)

492736

Sigma-Aldrich

Triphenylarsine

99.99% trace metals basis

Synonym(s):
Arsinetriphenyl
Linear Formula:
(C6H5)3As
CAS Number:
Molecular Weight:
306.23
EC Number:
MDL number:
PubChem Substance ID:

assay

99.99% trace metals basis

form

solid

reaction suitability

reagent type: catalyst

mp

58-61 °C (lit.)

SMILES string

c1ccc(cc1)[As](c2ccccc2)c3ccccc3

InChI

1S/C18H15As/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H

InChI key

BPLUKJNHPBNVQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 33 Arsenic

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Stanisław Popiel et al.
Analytica chimica acta, 837, 52-63 (2014-07-09)
Novel solid-phase microextraction (SPME) fibres containing methyl, ethyl, butyl acrylate and methacrylate were first prepared by a sol-gel technique and investigated for determination of selected organoarsenic compounds (lewisite, methyldichloroarsine, phenyldichloroarsine, diphenylchloroarsine and triphenylarsine) from water samples. The influence of sorption
Veronika Sele et al.
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS), 30, 171-179 (2015-01-27)
In the present study liver samples (n=26) of Northeast Arctic cod (Gadus morhua), ranging in total arsenic concentrations from 2.1 to 240mg/kg liver wet weight (ww), were analysed for their content of total arsenic and arsenic species in the lipid-soluble

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service