Skip to Content
MilliporeSigma

492892

Boc-Ala-OH-3-13C

99 atom % 13C

Synonym(s):

N-(tert-Butoxycarbonyl)-L-alanine-3-13C, L-Alanine-3-13C, N-t-Boc derivative

Sign In to View Organizational & Contract Pricing.
Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

About This Item

Linear Formula:
13CH3CH(NH-Boc)CO2H
CAS Number:
Molecular Weight:
190.20
UNSPSC Code:
12352202
PubChem Substance ID:
MDL number:
Isotopic purity:
99 atom % 13C
Mass shift:
M+1
Form:
solid

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI

1S/C8H15NO4/c1-5(6(10)11)9-7(12)13-8(2,3)4/h5H,1-4H3,(H,9,12)(H,10,11)/t5-/m0/s1/i1+1

SMILES string

CC(C)(C)OC(=O)N[C@@H]([13CH3])C(O)=O

InChI key

QVHJQCGUWFKTSE-BXMHHZGSSA-N

isotopic purity

99 atom % 13C

form

solid

optical activity

[α]20/D −23°, c = 2 in acetic acid

technique(s)

bio NMR: suitable

mp

79-83 °C (lit.)

application(s)

peptide synthesis

functional group

Boc

mass shift

M+1

Quality Level

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
586749486760485837
mass shift

M+1

mass shift

M+3

mass shift

M+1

mass shift

M+4

isotopic purity

99 atom % 13C

isotopic purity

99 atom % 13C

isotopic purity

99 atom % 13C

isotopic purity

99 atom % 13C, 98 atom % 15N

technique(s)

bio NMR: suitable

technique(s)

bio NMR: suitable

technique(s)

bio NMR: suitable

technique(s)

bio NMR: suitable

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

solid

form

solid

form

solid

form

solid

mp

79-83 °C (lit.)

mp

79-83 °C (lit.)

mp

79-83 °C (lit.)

mp

79-83 °C (lit.)

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service