Documents

494631

Sigma-Aldrich

Cesium titanate

≥99.9% trace metals basis

Synonym(s):
Cesium hexatitanate
Linear Formula:
Cs2Ti6O13
CAS Number:
Molecular Weight:
761.01
MDL number:

assay

≥99.9% trace metals basis

mp

>300 °C (lit.)

SMILES string

[Cs+].[Cs+].[O-][Ti](=O)O[Ti](=O)O[Ti](=O)O[Ti](=O)O[Ti](=O)O[Ti]([O-])=O

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service