Documents

495271

Sigma-Aldrich

(1S,4S)-(−)-2-(4-Methylphenyl)-2,5-diazabicyclo[2.2.1]heptane maleate salt

95%

Empirical Formula (Hill Notation):
C12H16N2 · C4H4O4
CAS Number:
Molecular Weight:
304.34
MDL number:
PubChem Substance ID:

assay

95%

optical activity

[α]20/D −72°, c = 1 in H2O

mp

174-178 °C (lit.)

SMILES string

OC(=O)\C=C/C(O)=O.Cc1ccc(cc1)N2C[C@@H]3C[C@H]2CN3

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service