Skip to Content
MilliporeSigma

497452

Dichloro[(S,S)-ethylenebis(4,5,6,7-tetrahydro-1-indenyl)]titanium(IV)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H24Cl2Ti
CAS Number:
Molecular Weight:
383.18
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352005
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


optical activity

[α]21/D −3486°, c = 0.07 in methylene chloride

mp

175-180 °C (lit.)

SMILES string

Cl[Ti]1(Cl)[C@@]4(CC[C@@]12C=CC3=C2CCCC3)C=CC5=C4CCCC5

InChI

1S/C20H24.2ClH.Ti/c1-3-7-19-15(5-1)9-11-17(19)13-14-18-12-10-16-6-2-4-8-20(16)18;;;/h9-12H,1-8,13-14H2;2*1H;/q;;;+2/p-2

InChI key

JCEDTJKHTKLRJD-UHFFFAOYSA-L



Pictograms

Corrosion

Signal Word

Danger

Hazard Codes

Hazard Classifications

Skin Corr. 1B

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library