499072

Sigma-Aldrich

4-Pentenylzinc bromide solution

0.5 M in THF

Linear Formula:
H2C=CH(CH2)3ZnBr
CAS Number:
Molecular Weight:
214.42
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

concentration

0.5 M in THF

density

0.954 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Br[Zn]CCCC=C

InChI

1S/C5H9.BrH.Zn/c1-3-5-4-2;;/h3H,1-2,4-5H2;1H;/q;;+1/p-1

InChI key

OQBZNYKGPFEIEK-UHFFFAOYSA-M

Related Categories

Packaging

50 mL in Sure/Seal™

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Supp Hazards

EUH018 - EUH019

RIDADR

UN 2924 8(3) / PGII

WGK Germany

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.