499366

Sigma-Aldrich

4-Isopropylphenylzinc iodide solution

0.5 M in THF

Linear Formula:
(CH3)2CHC6H4Znl
CAS Number:
Molecular Weight:
311.48
MDL number:
PubChem Substance ID:

concentration

0.5 M in THF

bp

65 °C

density

0.997 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(C)c1ccc([Zn]I)cc1

InChI

1S/C9H11.HI.Zn/c1-8(2)9-6-4-3-5-7-9;;/h4-8H,1-2H3;1H;/q;;+1/p-1

InChI key

IVLFHYKESLWRMK-UHFFFAOYSA-M

Related Categories

Packaging

50 mL in Sure/Seal™

Legal Information

Product of Rieke® Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Statements

Supp Hazards

May form explosive peroxides.

RIDADR

UN 2924 8(3) / PGII

WGK Germany

3

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.