510289

Sigma-Aldrich

Bismuth aluminate hydrate

≥99.9%

Synonym(s):
Aluminum bismuth oxide hydrate
Linear Formula:
Bi2(Al2O4)3 · xH2O
CAS Number:
Molecular Weight:
771.84 (anhydrous basis)
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥99.9%

reaction suitability

core: bismuth
reagent type: catalyst

SMILES string

[Bi+3].[O-][Al]=O.[O-][Al]=O.[O-][Al]=O

InChI

1S/3Al.Bi.6O/q;;;+3;;;;3*-1

InChI key

ZWSUTHPTOGDVHM-UHFFFAOYSA-N

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.