513482

Sigma-Aldrich

2,5-Bis(tert-butylperoxy)-2,5-dimethylhexane, blend with polypropylene and silica

Synonym(s):
Luperox® 101PP20
Linear Formula:
[(CH3)3COOC(CH3)2CH2-]2
CAS Number:
Molecular Weight:
290.44
MDL number:

concentration

~20 wt. % in polypropylene and silica

mp

146-153 °C (lit.)

SMILES string

CC(C)(C)OOC(C)(C)CCC(C)(C)OOC(C)(C)C

Application

Polymerization initiator

Legal Information

Product of Arkema Inc.
Luperox is a registered trademark of Arkema Inc.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service