All Photos(1)

513709

Sigma-Aldrich

1,1-Dimethyl-N-tert-octylpropargylamine

96%

Linear Formula:
HC≡CC(CH3)2NHC(CH3)2CH2C(CH3)3
CAS Number:
Molecular Weight:
195.34
MDL number:
PubChem Substance ID:

assay

96%

refractive index

n20/D 1.45 (lit.)

bp

206 °C (lit.)

density

0.815 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)CC(C)(C)NC(C)(C)C#C

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service