Documents

514829

Sigma-Aldrich

Bis[4-(vinyloxy)butyl] 1,6-hexanediylbiscarbamate

98%

Synonym(s):
VEctomer® 4230 vinyl ether
Linear Formula:
[(CH2)3NHCO2(CH2)4OCH=CH2]2
CAS Number:
Molecular Weight:
400.51
MDL number:
PubChem Substance ID:

assay

98%

mp

99-103 °C (lit.)

SMILES string

C=COCCCCOC(=O)NCCCCCCNC(=O)OCCCCOC=C

InChI

1S/C20H36N2O6/c1-3-25-15-9-11-17-27-19(23)21-13-7-5-6-8-14-22-20(24)28-18-12-10-16-26-4-2/h3-4H,1-2,5-18H2,(H,21,23)(H,22,24)

InChI key

SQPFTUPRJYKNIL-UHFFFAOYSA-N

Legal Information

Product of Morflex, Inc.
VEctomer is a registered trademark of Vertellus Specialties, Inc.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service