All Photos(1)

517593

Sigma-Aldrich

Praseodymium(III) hexafluoroacetylacetonate hydrate

≥99.5%

Linear Formula:
Pr(C5HF6O2)3 · xH2O
CAS Number:
Molecular Weight:
762.06 (anhydrous basis)
MDL number:
PubChem Substance ID:

assay

≥99.5%

form

solid

reaction suitability

core: praseodymium
reagent type: catalyst

mp

114.5-127.7 °C (lit.)

SMILES string

O.FC(F)(F)C(=O)\C=C(/O[Pr](O\C(=C/C(=O)C(F)(F)F)C(F)(F)F)O\C(=C/C(=O)C(F)(F)F)C(F)(F)F)C(F)(F)F

InChI

1S/3C5H2F6O2.H2O.Pr/c3*6-4(7,8)2(12)1-3(13)5(9,10)11;;/h3*1,12H;1H2;/q;;;;+3/p-3/b3*2-1-;;

InChI key

KNKOGZDXDBKNGE-AHUNZLEGSA-K

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service